Anti-TRI18 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB128363-200EA 0 DKK
BIRBORB128363-200 BIRBORB128363-100
Anti-TRI18 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRI18
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR