Anti-PTK2 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB127424-100EA 0 DKK
BIRBORB127424-100 BIRBORB127424-200
Anti-PTK2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat