Anti-KISS1R Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB124715-100EA 0 DKK
BIRBORB124715-100
Anti-KISS1R Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: KISS1R (KISS1 receptor)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR