Anti-F-Actin Rabbit Polyclonal Antibody (Cy5.5®)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB123142-100EA 0 DKK
BIRBORB123142-100
Anti-F-Actin Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
Type: Primary
Antigen: F-Actin
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR