Anti-PYCARD Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB113899-100EA 0 DKK
BIRBORB113899-100
Anti-PYCARD Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Type: Primary
Antigen: PYCARD (PYD and CARD domain containing)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR