Anti-FLT3LG Rabbit polyclonal antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB66818-100EA 0 DKK
BIRBORB66818-100
Anti-FLT3LG Rabbit polyclonal antibody
Antistoffer
Western Blot: 1:1000-1:5000; ELISA: 1:1000-1:10000

Type: Primary
Antigen: FLT3LG (fms-related tyrosine kinase 3 ligand)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR