Anti-PCNA Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48817-200EA 0 DKK
BIRBORB48817-200
Anti-PCNA Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:2000 (Sensitivity 500 ng/ml)

Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR