Anti-PIK3C3 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48746-200EA 0 DKK
BIRBORB48746-200
Anti-PIK3C3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PIK3C3 (phosphatidylinositol 3-kinase, catalytic subunit type 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR