Anti-BIScV Chicken Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48710-200EA 0 DKK
BIRBORB48710-200
Anti-BIScV Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BIScV
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Virus
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR