Anti-MAPT Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48689-200EA 0 DKK
BIRBORB48689-200
Anti-MAPT Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR