Anti-SMOC1 Chicken Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48623-200EA 0 DKK
BIRBORB48623-200
Anti-SMOC1 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SMOC1 (SPARC related modular calcium binding 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR