Anti-BTRC Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48295-100EA 0 DKK
BIRBORB48295-100
Anti-BTRC Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BTRC (beta-transducin repeat containing E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR