Anti-BMPR1B Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48291-100EA 0 DKK
BIRBORB48291-100
Anti-BMPR1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BMPR1B (bone morphogenetic protein receptor, type IB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR