Anti-His tag Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48087-100EA 0 DKK
BIRBORB48087-100
Anti-His tag Mouse Monoclonal Antibody
Antistoffer
  • Western Blot: 1:5000; Immunofluorescence: 1:500-1:2000; Immunochemistry: 1:100

Type: Primary
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR