Anti-CDK2 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48014-100EA 0 DKK
BIRBORB48014-100
Anti-CDK2 Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR