Anti-COL4A1 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB48003-100EA 0 DKK
BIRBORB48003-100
Anti-COL4A1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COL4A1 (collagen, type IV, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR