Anti-Caspase 4 p10 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB47957-100EA 2570 DKK
BIRBORB47957-100
Anti-Caspase 4 p10 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Caspase 4 p10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human