Anti-AQP3 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB47955-100EA 0 DKK
BIRBORB47955-100
Anti-AQP3 Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: AQP3 (aquaporin 3 (Gill blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR