Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB44818-0.1EA 0 DKK
BIRBORB44818-0.1
Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR