Anti-CD9 Rat Monoclonal Antibody (PE (Phycoerythrin))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB44806-0.1EA 0 DKK
BIRBORB44806-0.1
Anti-CD9 Rat Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR