Anti-LAMP1 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB44716-100EA 0 DKK
BIRBORB44716-100
Anti-LAMP1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR