Anti-CDH1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB44674-100EA 0 DKK
BIRBORB44674-100
Anti-CDH1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR