Anti-CD40LG Hamster Monoclonal Antibody (FITC (Fluorescein))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB44603-0.1EA 0 DKK
BIRBORB44603-0.1
Anti-CD40LG Hamster Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR