Anti-SELPLG Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB44511-0.1EA 0 DKK
BIRBORB44511-0.1
Anti-SELPLG Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: SELPLG (selectin P ligand)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human