Anti-ITGA2B Mouse Monoclonal Antibody (FITC (Fluorescein))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43998-100EA 0 DKK
BIRBORB43998-100
Anti-ITGA2B Mouse Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR