Anti-CD22 Mouse Monoclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43980-0.1EA 0 DKK
BIRBORB43980-0.1
Anti-CD22 Mouse Monoclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR