Anti-Hsp90 beta Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43978-0.1EA 0 DKK
BIRBORB43978-0.1
Anti-Hsp90 beta Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Hsp90 beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR