Anti-CD81 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43974-100EA 0 DKK
BIRBORB43974-100
Anti-CD81 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR