Anti-Chorionic gonadotropin Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43808-0.1EA 2490 DKK
BIRBORB43808-0.1
Anti-Chorionic gonadotropin Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: Chorionic gonadotropin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR