Anti-TNFRSF21 Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB43760-0.1EA 0 DKK
BIRBORB43760-0.1
Anti-TNFRSF21 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: TNFRSF21 (tumor necrosis factor receptor superfamily, member 21)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human