Anti-CD28 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43728-100EA 0 DKK
BIRBORB43728-100
Anti-CD28 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Antistoffer
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR