Anti-beta endorphin Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43665-0.1EA 0 DKK
BIRBORB43665-0.1
Anti-beta endorphin Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: beta endorphin (beta endorphin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR