Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43608-100EA 0 DKK
BIRBORB43608-100
Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antistoffer
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR