Anti-DDX4 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43493-100EA 0 DKK
BIRBORB43493-100
Anti-DDX4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DDX4 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR