Anti-CRABP2 Rabbit Polyclonal Antibody
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB43484-100EA 0 DKK
BIRBORB43484-100
Anti-CRABP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu