Anti-SLC30A6 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43311-100EA 0 DKK
BIRBORB43311-100
Anti-SLC30A6 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:500

Type: Primary
Antigen: SLC30A6 (solute carrier family 30 (zinc transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR