Anti-KRT18 Mouse Monoclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB43292-100EA 0 DKK
BIRBORB43292-100
Anti-KRT18 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR