Anti-CERS2 Rabbit polyclonal antibody
Leverandør: Biorbyt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB43287-100EA 0 DKK
BIRBORB43287-100
Anti-CERS2 Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: CERS2 (ceramide synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human