Anti-CUL7 Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB41832-100EA 0 DKK
BIRBORB41832-100
Anti-CUL7 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:500-1:3000

Type: Primary
Antigen: CUL7 (cullin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR