Anti-KPNA4 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB41246-100EA 0 DKK
BIRBORB41246-100
Anti-KPNA4 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:500-1:3000 Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: KPNA4 (karyopherin alpha 4 (importin alpha 3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR