Anti-ARHGEF18 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB40915-100EA 0 DKK
BIRBORB40915-100
Anti-ARHGEF18 Rabbit Polyclonal Antibody
Antistoffer
  • Western Blot: 1:500-1:3000

Type: Primary
Antigen: ARHGEF18 (Rho/Rac guanine nucleotide exchange factor (GEF) 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR