Anti-APPL1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27763-100EA 0 DKK
BIRBORB27763-100
Anti-APPL1 Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: APPL1 (adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR