Anti-IgG Rabbit Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27728-0.5EA 0 DKK
BIRBORB27728-0.5 BIRBORB27728-1
Anti-IgG Rabbit Antibody
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR