Anti-IL23A Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB27657-100EA 0 DKK
BIRBORB27657-100
Anti-IL23A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL23A (interleukin 23, alpha subunit p19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse