Anti-IGF1 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27586-100EA 0 DKK
BIRBORB27586-100
Anti-IGF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IGF1 (insulin-like growth factor 1 (somatomedin C))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR