Anti-CEL Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27495-50EA 0 DKK
BIRBORB27495-50
Anti-CEL Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: CEL (carboxyl ester lipase)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR