Anti-AGE:BIO Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27491-20EA 0 DKK
BIRBORB27491-20
Anti-AGE:BIO Mouse Monoclonal Antibody
Antistoffer
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: AGE:BIO
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR