Anti-Fibronectin Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB27336-0.5EA 0 DKK
BIRBORB27336-0.5 BIRBORB27336-0.1
Anti-Fibronectin Rabbit polyclonal antibody
Antistoffer
ImmunofluorescenceA 1:80; Immunochemistry: 1:500; ELISA: 1:200

Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR