Anti-Epithelial Membrane Antigen Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB27219-5EA 0 DKK
BIRBORB27219-5
Anti-Epithelial Membrane Antigen Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Epithelial Membrane Antigen
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human