Anti-COL4 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26867-0.5EA 0 DKK
BIRBORB26867-0.5
Anti-COL4 Rabbit Polyclonal Antibody
Antistoffer
Immunofluorescence: 1:40; Immunochemistry: 1:250

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR